Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hydroxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2kg
500g
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

1305-62-0
CaH2O2
74.092
AXCZMVOFGPJBDE-UHFFFAOYSA-L
6093208
CHEBI:31341

High quality chemicals for laboratory and specialized industrial use

CAS 1305-62-0
Molecular Formula CaH2O2
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca+2]
IUPAC Name calcium;dihydroxide
Molecular Weight (g/mol) 74.092
PubChem CID 6093208
ChEBI CHEBI:31341
Formula Weight 74.09
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 2.24kg
Quantity 500g
Chemical Name or Material Calcium Hydroxide
Grade Baker Analyzed™ ACS Reagent
Show More Show Less