Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hydroxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

1305-62-0
CaH2O2
74.092
AXCZMVOFGPJBDE-UHFFFAOYSA-L
6093208
CHEBI:31341
calcium;dihydroxide
[OH-].[OH-].[Ca+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 74.09
Physical Form Powder
Packaging Poly Pail
Specific Gravity 1L = 2.24kg
Quantity 2kg
Chemical Name or Material Calcium Hydroxide
Grade Baker Analyzed™ ACS Reagent