Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hydroxide, Powder, FCC, 95-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1305-62-0
CaH2O2
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium dihydroxide
[OH-].[OH-].[Ca++]
1305-62-0
CaH2O2
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium dihydroxide
[OH-].[OH-].[Ca++]

C1116, 1305-62-0, Ca(OH)2

Spectrum™ Calcium Hydroxide, Powder, FCC is used extensively in the food industry to clarify solutions, as a digestion aid, baking soda substitute and as a nutritional supplement because of its low toxicity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 95-100.5%
Fluoride 0.005%
Insoluble in Acid 0.5%
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Quantity 2.5 kg