Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hypochlorite, Granular, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
7778-54-3
7778-54-3
Grade Reagent
CAS Max % 1
Available Chlorine Approx. 65%
Packaging Poly Pail
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.