Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hypochlorite, Purified, 65% minimum Available Chlorine, Granular, Purified, J.T.Baker™

Click to view available options
Quantity:
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7778-54-3
CaCl2O2
142.976
ZKQDCIXGCQPQNV-UHFFFAOYSA-N
24504
calcium;dihypochlorite
[O-]Cl.[O-]Cl.[Ca+2]
Formula Weight 142.98
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 2.35kg
Quantity 2.5kg
Chemical Name or Material Calcium Hypochlorite, Purified
Assay ≥65% available chlorine
Grade Purified