Calcium Hypochlorite, Purified, 65% minimum Available Chlorine, Granular, Purified, J.T.Baker™

Manufacturer: J.T.Baker 137801

Packaging Wide Mouth Amber Glass Bottle
Quantity 500g

 View more versions of this product

Catalog No. 02-002-533

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7778-54-3
    Molecular Formula CaCl2O2
    Formula Weight 142.98
    Chemical Name or Material Calcium Hypochlorite, Purified
    Grade Purified

    Specifications

    Formula Weight 142.98
    Packaging Wide Mouth Amber Glass Bottle
    Specific Gravity 1L = 2.35kg
    Molecular Formula CaCl2O2
    CAS 7778-54-3
    Quantity 500g
    Physical Form Granular
    Chemical Name or Material Calcium Hypochlorite, Purified
    Assay ≥65% available chlorine
    Grade Purified

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit