Calcium hypochlorite, tech., Alfa Aesar™

For Use in Deodorant, Oxidizing Agent, Bleaching Agent

Manufacturer: Alfa Aesar 4254809

Quantity 10g

 View more versions of this product

Catalog No. AA4254809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7778-54-3
    Physical Form Granular
    Molecular Formula CaCl2O2
    Formula Weight 142.98
    MDL Number MFCD00010900
    Synonym Bleaching powder
    Chemical Name or Material Calcium hypochlorite

    Specifications

    Formula Weight 142.98
    Quantity 10g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    Density 2.35g/cm3 at 20°C
    EINECS Number 231-908-7
    Physical Form Granular
    Molecular Formula CaCl2O2
    MDL Number MFCD00010900
    CAS 7778-54-3
    Synonym Bleaching powder
    Health Hazard 1 H272-H314-H400-H302 : May intensify fire; oxidizer. Causes severe skin burns and eye damage. Very toxic to aquatic life. Harmful if swallowed.
    Appearance White granular powder
    Chemical Name or Material Calcium hypochlorite

    H272-H314-H400-H302 : May intensify fire; oxidizer. Causes severe skin burns and eye damage. Very toxic to aquatic life. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit