Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium iodate monohydrate, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4013822

Quantity 100g

 View more versions of this product

Catalog No. AA4013822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10031-32-0
>99%
407.90 (389.88 Anhydrous)
Calcium iodate monohydrate
Powder
Ca(IO3)2·H2O
MFCD00036265

Specifications

10031-32-0
>99%
Off-white powder
MFCD00036265
100g
Powder
Ca(IO3)2·H2O
407.90 (389.88 Anhydrous)
Off-white
Calcium iodate monohydrate

Safety and Handling

SDS & Safety Documents

H272-H315-H319-H335 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 232-191-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit