Calcium iodide, ultra dry, 99.99% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer: Alfa Aesar 4367414

Quantity 25g

 View more versions of this product

Catalog No. AA4367414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-68-8
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula CaI2
    Formula Weight 293.89
    MDL Number MFCD00010910
    Chemical Name or Material Calcium iodide

    Specifications

    Formula Weight 293.89
    Quantity 25g
    Color Off-white to pale yellow
    Density 3.95g/cm3 at 20°C
    EINECS Number 233-276-8
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula CaI2
    Appearance Off-white to pale yellow powder or beads
    MDL Number MFCD00010910
    CAS 10102-68-8
    Health Hazard 1 H315-H319 : Causes skin irritation. Causes serious eye irritation.
    Chemical Name or Material Calcium iodide

    H315-H319 : Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit