Alfa Aesar™ Calcium iodide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4425414

Quantity 25g

 View more versions of this product

Catalog No. AA4425414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-68-8
    Molecular Formula CaI2
    Formula Weight 293.89
    MDL Number MFCD00010910
    Merck Index 14,1677
    Chemical Name or Material Calcium iodide, Ultra dry

    Specifications

    Formula Weight 293.89
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Mesh Size −10 Mesh
    Physical Form Beads
    CAS 10102-68-8
    Merck Index 14,1677
    Boiling Point 1100°C
    Density 3.956g/mL
    TSCA TSCA
    Odor Odorless
    Sensitivity Light Sensitive & Hygroscopic
    Molecular Formula CaI2
    MDL Number MFCD00010910
    Color White to pale yellow
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Storage Note 1
    • Ampouled under argon
    Melting Point 784°C
    Chemical Name or Material Calcium iodide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit