Learn More
Calcium Lactate, Monohydrate, USP, 98-101%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Pail
CA153, 41372-22-9, C6H10O6Ca•H2O
Spectrum™ Calcium Lactate, Monohydrate, USP is used as an antacid and mineral supplement to treat calcium deficiencies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Acidity 0.0045
Heavy Metals 2e-005
Loss on Drying 5.0-8%
Packaging Fiber Drum