Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Lactate, Monohydrate, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
10 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
41372-22-9
C6H12CaO7
236.23
MDAVASCOAJMZHZ-UHFFFAOYNA-L
calcium bis(2-hydroxypropanoate) hydrate
O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
41372-22-9
C6H12CaO7
236.23
MDAVASCOAJMZHZ-UHFFFAOYNA-L
calcium bis(2-hydroxypropanoate) hydrate
O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O

CA153, 41372-22-9, C6H10O6Ca•H2O

Spectrum™ Calcium Lactate, Monohydrate, USP is used as an antacid and mineral supplement to treat calcium deficiencies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Acidity 0.45%
CAS Max % 100%
Percent Purity 98-101%
Heavy Metals 0.002%
Loss on Drying 5.0-8%
Packaging Poly Pail
Quantity 10 kg