Alfa Aesar™ Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 1068322

Quantity 100g

 View more versions of this product

Catalog No. AA1068322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35054-52-5
    Formula Weight 164.09 (Anhydrous)
    MDL Number MFCD00149604
    UN Number UN1454
    Merck Index 14,1682
    Chemical Name or Material Calcium nitrate hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 164.09 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00149604
    UN Number UN1454
    Melting Point 42°C
    Health Hazard 2 GHS H Statement
    H272-H302-H315-H319-H335
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 1.82g/mL
    Physical Form Crystalline Aggregates
    Sensitivity Hygroscopic
    Merck Index 14,1682
    Boiling Point 132°C (dec.)
    Health Hazard 1 Danger
    CAS 35054-52-5
    Quantity 100g
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
    RTECS Number EW3000000
    Chemical Name or Material Calcium nitrate hydrate
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H272-H302-H315-H319-H335
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit