Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Nitrate Tetrahydrate, ACS Reagent, Reagents
Click to view available options
Quantity:
125 g
500 g
50 lb.
50 kg
Specifications
Specifications
| Boiling Point | 132°C |
| Melting Point | 561°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 50 lb. |
| Grade | ACS |
| Packaging | Fiber Drum |
| Specific Gravity | 1.82 |
| Chemical Name or Material | Calcium Nitrate Tetrahydrate, ACS Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction