Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Nitrate Tetrahydrate, Honeywell Fluka™

Click to view available options
Quantity:
250g
500g
1kg
5kg
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate,calcium nitrate hydrate, puratronic,acmc-1aekc,calcium nitrate tetra hydrate,ca.2no3.4h2o,ca no3 2.4h2o,calcium tetrahydrate dinitronate,calcium nitrate-water 1/4,calcium tetrahydrate dinitrate,calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate,calcium nitrate hydrate, puratronic,acmc-1aekc,calcium nitrate tetra hydrate,ca.2no3.4h2o,ca no3 2.4h2o,calcium tetrahydrate dinitronate,calcium nitrate-water 1/4,calcium tetrahydrate dinitrate,calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]

Puriss. p.a., ACS Reagent, 99-103%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula Ca(NO3)2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 236.15g/mol
Percent Purity 99 to 103%
Grade ACS/puriss. p.a.
Quantity 250g
Chemical Name or Material Calcium Nitrate Tetrahydrate

P280-P305 + P351 + P338

H302-H318

EINECSNumber : 233-332-1