Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium oxalate monohydrate, 99.9985% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1300706

Quantity 5g

 View more versions of this product

Catalog No. AA1300706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

5794-28-5
99.9985% (Metal basis)
146.11
Calcium oxalate monohydrate, H2O at 200°
Powder
CaC2O4·H2O
MFCD00150039

Specifications

5794-28-5
99.9985% (Metal basis)
White powder
MFCD00150039
Odorless
5g
Powder
CaC2O4·H2O
146.11
2.12g/cm3
White
Calcium oxalate monohydrate, H2O at 200°

Safety and Handling

SDS & Safety Documents

H302-H312

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 209-260-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit