Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Calcium oxalate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4631814

Quantity 25g

 View more versions of this product

Catalog No. AA4631814


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

563-72-4
CaC2O4
MFCD00012474
Calcium oxalate
C2CaO4
128.1
14,1685
Puratronic™

Specifications

563-72-4
C2CaO4
128.1
14,1685
Hygroscopic
25g
99.999%, (metals basis)
Powder
CaC2O4
MFCD00012474
2.2g/mL
White
Calcium oxalate
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit