Alfa Aesar™ Calcium oxalate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4631806

Quantity 5g

 View more versions of this product

Catalog No. AA4631806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 563-72-4
    Molecular Formula C2CaO4
    Linear Formula CaC2O4
    Formula Weight 128.1
    MDL Number MFCD00012474
    Merck Index 14,1685
    Chemical Name or Material Calcium oxalate
    Grade Puratronic™

    Specifications

    Formula Weight 128.1
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    CAS 563-72-4
    TSCA TSCA
    Sensitivity Hygroscopic
    Linear Formula CaC2O4
    MDL Number MFCD00012474
    Molecular Formula C2CaO4
    Merck Index 14,1685
    Color White
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.
    Density 2.2g/mL
    Health Hazard 1 Danger
    Chemical Name or Material Calcium oxalate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.

    GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit