Calcium oxide, Puratronic, 99.998% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 1068418

Quantity 50g

 View more versions of this product

Catalog No. AA1068418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1305-78-8
    Physical Form Powder
    Assay Percent Range 99.99% (metals basis, excluding other alkaline earth and alkali metals 130ppm max.)
    Molecular Formula CaO
    Formula Weight 56.08
    MDL Number MFCD00010911
    Chemical Name or Material Calcium oxide

    Specifications

    Formula Weight 56.08
    Quantity 50g
    Color White
    EINECS Number 215-138-9
    Health Hazard 1 H314-H318
    Storage Note 1 Air & Moisture Sensitive
    DOT Information DOT Class: 8, Packaging Group: III
    Physical Form Powder
    Density 3.25-3.38
    Assay Percent Range 99.99% (metals basis, excluding other alkaline earth and alkali metals 130ppm max.)
    Molecular Formula CaO
    MDL Number MFCD00010911
    CAS 1305-78-8
    Chemical Name or Material Calcium oxide

    Air & Moisture Sensitive

    H314-H318

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit