Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Powder, Technical, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
1305-78-8
CaO
56.08
ODINCKMPIJJUCX-UHFFFAOYSA-N
oxocalcium
O=[Ca]
1305-78-8
CaO
56.08
ODINCKMPIJJUCX-UHFFFAOYSA-N
oxocalcium
O=[Ca]

C1141, 1305-78-8, CaO

Spectrum™ Calcium Oxide, Powder, Technical , is a widely used chemical compound known as quicklime. A white, alkaline crystalline solid that is also caustic. It is a component of common cement and used in the fabrication of paper and plaster. In the petroleum industry, calcium oxide and phenolpthalein are combined into a water detection paste .
CAS Max % 100%
UN Number UN1910
Packaging Poly Bottle
Quantity 500 g