Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Pantothenate, Powder, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
137-08-6
C18H32CaN2O10
476.54
FAPWYRCQGJNNSJ-UHFFFAOYNA-L
calcium bis(3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate)
[Ca++].CC(C)(CO)C(O)C(=O)NCCC([O-])=O.CC(C)(CO)C(O)C(=O)NCCC([O-])=O
137-08-6
C18H32CaN2O10
476.54
FAPWYRCQGJNNSJ-UHFFFAOYNA-L
calcium bis(3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate)
[Ca++].CC(C)(CO)C(O)C(=O)NCCC([O-])=O.CC(C)(CO)C(O)C(=O)NCCC([O-])=O

CA159, 137-08-6, C18H32CaN2O10

Spectrum™ Calcium Pantothenate, Powder, USP is often used in dietary supplements a more stable of pantothenic acid (Vitamin B5) in the digestive tract. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98-102%
Loss on Drying 5%
Packaging Poly Pail
Quantity 12 kg
Optical Rotation +25.0° to +27.5°