Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Pantothenate, U.S.P., J.T.Baker™

Click to view available options
Quantity:
25g
1kg
Packaging:
Wide Mouth Amber Glass Bottle
137-08-6
C18H34CaN2O10
478.552
WIWQIMNDNVAWQN-UBKPKTQASA-N
calcium pantothenate,d-pantothenic acid hemicalcium salt,d-pantothenic acid, calcium salt,pantothenic acid, calcium salt, d,r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt,beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
131847364
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
CC(C)(CO)C(C(=O)NCCC(=O)O)O.CC(C)(CO)C(C(=O)NCCC(=O)O)O.[Ca]
137-08-6
C18H34CaN2O10
478.552
WIWQIMNDNVAWQN-UBKPKTQASA-N
calcium pantothenate,d-pantothenic acid hemicalcium salt,d-pantothenic acid, calcium salt,pantothenic acid, calcium salt, d,r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt,beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
131847364
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
CC(C)(CO)C(C(=O)NCCC(=O)O)O.CC(C)(CO)C(C(=O)NCCC(=O)O)O.[Ca]
Formula Weight 476.54
Grade USP
Packaging Wide Mouth Amber Glass Bottle
Quantity 1kg
Chemical Name or Material Calcium Pantothenate