Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, 96%, pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
7758-87-4
Ca3O8P2
310.18
QORWJWZARLRLPR-UHFFFAOYSA-H
calcium phosphate,synthos,tricalcium phosphate,tricalcium diphosphate,calcigenol simple,calcium orthophosphate,beta-tcp,tribasic calcium phosphate,bonarka,calcium phosphate, tribasic
24456
CHEBI:9679
tricalcium;diphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]
7758-87-4
Ca3O8P2
310.18
QORWJWZARLRLPR-UHFFFAOYSA-H
calcium phosphate,synthos,tricalcium phosphate,tricalcium diphosphate,calcigenol simple,calcium orthophosphate,beta-tcp,tribasic calcium phosphate,bonarka,calcium phosphate, tribasic
24456
CHEBI:9679
tricalcium;diphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]
Assay Percent Range 95% min. (Complexometry)
Linear Formula Ca3(PO4)2
Merck Index 15,1696
Solubility Information Solubility in water: 0.1g/L (25°C)
Formula Weight 310.18
Percent Purity 96%
Grade Pure
Packaging Glass bottle
Melting Point 1670.0°C
Quantity 1kg
Chemical Name or Material Calcium phosphate
Show More Show Less

EINECSNumber : 231-840-8