Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous, Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate,calcium hydrogenphosphate,calcium phosphate dibasic,calcium phosphate, dibasic,biofos,fujicalin s,dicafos an,dibasic calcium phosphate,monocalcium acid phosphate,phosphoric acid, calcium salt 1:1
24441
CHEBI:32596
calcium;hydrogen phosphate
OP(=O)([O-])[O-].[Ca+2]
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate,calcium hydrogenphosphate,calcium phosphate dibasic,calcium phosphate, dibasic,biofos,fujicalin s,dicafos an,dibasic calcium phosphate,monocalcium acid phosphate,phosphoric acid, calcium salt 1:1
24441
CHEBI:32596
calcium;hydrogen phosphate
OP(=O)([O-])[O-].[Ca+2]
Biological buffer
Assay Percent Range 98.0 to 103% (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Percent Purity 98+%
Grade Analysis
Chloride 0.033% max
Insoluble Matter 0.2% max. (in HCl)
Packaging Plastic bottle
Solubility Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Sulfate 0.5% max
Quantity 50g
Chemical Name or Material Calcium phosphate, dibasic
Show More Show Less

EINECSNumber : 231-826-1