Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium phosphate, dibasic, 97%, pure, anhydrous, ACROS Organics™
Click to view available options
Quantity:
25g
1kg
5kg
Packaging:
Plastic bottle
Specifications
Assay Percent Range | 96% min. (Complexometry) |
Linear Formula | CaHPO4 |
Merck Index | 15,1694 |
Solubility Information | Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol |
Formula Weight | 136.06 |
Physical Form | Powder |
Percent Purity | 97% |
Grade | Pure |
Packaging | Plastic bottle |
Solubility | Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol |
Show More |
Safety and Handling
EINECSNumber : 231-826-1