Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, dibasic, 97%, pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
1kg
5kg
Packaging:
Plastic bottle
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate,calcium hydrogenphosphate,calcium phosphate dibasic,calcium phosphate, dibasic,biofos,fujicalin s,dicafos an,dibasic calcium phosphate,monocalcium acid phosphate,phosphoric acid, calcium salt 1:1
24441
CHEBI:32596
calcium;hydrogen phosphate
OP(=O)([O-])[O-].[Ca+2]
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate,calcium hydrogenphosphate,calcium phosphate dibasic,calcium phosphate, dibasic,biofos,fujicalin s,dicafos an,dibasic calcium phosphate,monocalcium acid phosphate,phosphoric acid, calcium salt 1:1
24441
CHEBI:32596
calcium;hydrogen phosphate
OP(=O)([O-])[O-].[Ca+2]
Assay Percent Range 96% min. (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Physical Form Powder
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Solubility Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Color White
Quantity 25g
Chemical Name or Material Calcium phosphate, dibasic
Show More Show Less

EINECSNumber : 231-826-1