Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic, Anhydrous, Powder, 98%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7757-93-9
7757-93-9
Grade Reagent
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 0.98
Packaging Poly Pail
Quantity 12 kg

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.