Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic, Anhydrous, Powder, FCC, EP, BP, 98-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate
[Ca++].OP([O-])([O-])=O
7757-93-9
CaHO4P
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium hydrogen phosphate
[Ca++].OP([O-])([O-])=O

C1151, 7757-93-9, CaHPO4

Spectrum™ Calcium Phosphate Dibasic, Anhydrous, Powder, FCC, EP, BP is used as a calcium dietary supplement in prepared breakfast cereals, dog treats, enriched flour, and noodle products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 98-101%
Chloride 330 ppm
Fluoride 0.005%
Lead (Pb) 2 mg/kg
Loss on Ignition 7.0-8.5%
Packaging Fiber Drum
Sulfate 0.5%
Quantity 45 kg