Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, dibasic dihydrate, 98+%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
7789-77-7
CaHO42H2O
172.09
MFCD00149621
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium hydrogenphosphate dihydrate,calcium hydrogen phosphate dihydrate,unii-o7tsz97gep,calcium monohydrogen phosphate dihydrate,o7tsz97gep,calcium phosphate dibasic dihydrate,dibasic calcium phosphate dihydrate,phosphoric acid, calcium salt 1:1 , dihydrate,brushite,aksepillen
104805
CHEBI:4496
calcium;hydrogen phosphate;dihydrate
O.O.OP(=O)([O-])[O-].[Ca+2]
7789-77-7
CaHO42H2O
172.09
MFCD00149621
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium hydrogenphosphate dihydrate,calcium hydrogen phosphate dihydrate,unii-o7tsz97gep,calcium monohydrogen phosphate dihydrate,o7tsz97gep,calcium phosphate dibasic dihydrate,dibasic calcium phosphate dihydrate,phosphoric acid, calcium salt 1:1 , dihydrate,brushite,aksepillen
104805
CHEBI:4496
calcium;hydrogen phosphate;dihydrate
O.O.OP(=O)([O-])[O-].[Ca+2]
Assay Percent Range 98.0 to 105% (on dried substance) (Complexometry)
Linear Formula CaHPO4·2H2O
Solubility Information Solubility in water: 0.2g/L (25°C)
Formula Weight 172.09
Physical Form Powder
Percent Purity 98+%
Grade Extra Pure
Identification Calcium: positive, Phosphate: positive
Loss on Ignition 24.5% to 26.5% (800°C)
Packaging Plastic bottle
Solubility Solubility in water: 0.2g/L (25°C).
Color White
Quantity 1kg
Chemical Name or Material Calcium phosphate, dibasic dihydrate
Show More Show Less

GHS Signal Word: Warning