Learn More
Calcium Phosphate Dibasic, Dihydrate, Powder, FCC, 97-105%, Spectrum™
Click to view available options
Packaging:
Poly Bottle
Poly Pail
C1809, 7789-77-7, CaHPO4•2H2O
Spectrum™ Calcium Phosphate Dibasic, Dihydrate, Powder, FCC is used as a calcium dietary supplement in prepared breakfast cereals, dog treats, enriched flour, and noodle products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Arsenic (As) 3 mg/kg
Lead (Pb) 2 mg/kg
Packaging Poly Bottle