Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic, Dihydrate, Powder, FCC, 97-105%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
7789-77-7
CaH5O6P
172.09
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium dihydrate hydrogen phosphate
O.O.[Ca++].OP([O-])([O-])=O
7789-77-7
CaH5O6P
172.09
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium dihydrate hydrogen phosphate
O.O.[Ca++].OP([O-])([O-])=O

C1809, 7789-77-7, CaHPO4•2H2O

Spectrum™ Calcium Phosphate Dibasic, Dihydrate, Powder, FCC is used as a calcium dietary supplement in prepared breakfast cereals, dog treats, enriched flour, and noodle products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 97-105%
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Quantity 2.5 kg