Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC, 15.9-17.7%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10031-30-8
CaH2O9P2
248.04
ZBZJARSYCHAEND-UHFFFAOYSA-H
calcium hydrate diphosphate
O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
10031-30-8
CaH2O9P2
248.04
ZBZJARSYCHAEND-UHFFFAOYSA-H
calcium hydrate diphosphate
O.[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

CA158, 10031-30-8, Ca(H2PO4)2•H2O

Spectrum™ Calcium Phosphate, Monobasic, Monohydrate, Powder, FCC is used in the food industry as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 15.9-17.7%
Lead (Pb) 2 mg/kg
Loss on Drying 1%
Packaging Poly Bottle
Quantity 500 g