Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  J.T.Baker 143001

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Catalog No. 02-002-698

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7757-93-9
136.06
BAKER ANALYZED™ Reagent
CaHPO4
Calcium Phosphate

Specifications

7757-93-9
CaHPO4
Poly Bottle
500g
BAKER ANALYZED™ Reagent
Powder
136.06
1L = 2.30kg
Calcium Phosphate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit