Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate, Tribasic, Powder, FCC, 34-40%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1306-06-5
Ca5HO13P3
502.31
XYJRXVWERLGGKC-UHFFFAOYSA-D
pentacalcium hydroxide triphosphate
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1306-06-5
Ca5HO13P3
502.31
XYJRXVWERLGGKC-UHFFFAOYSA-D
pentacalcium hydroxide triphosphate
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

C1144, 1306-06-5, Ca5OH(PO4)3

Spectrum™ Calcium Phosphate, Tribasic, Powder, FCC is used in powdered spices as an anticaking agent, a raising agent, an antacid and nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 mg/kg
Percent Purity 34-40%
Fluoride 0.0075%
Lead (Pb) 2 mg/kg
Loss on Ignition 10%
Packaging Fiber Drum
Quantity 12 kg