Calcium phosphide, Red-brown lump, Alfa Aesar™

Manufacturer: Alfa Aesar 1911514

Quantity 25g

 View more versions of this product

Catalog No. AA1911514

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1305-99-3
    Physical Form Lump
    Molecular Formula Ca3P2
    Formula Weight 182.19
    MDL Number MFCD00015986
    Chemical Name or Material Calcium phosphide
    Grade Technical Grade

    Specifications

    Physical Form Lump
    Formula Weight 182.19
    Quantity 25g
    Color Red-brown
    DOT Information DOT Class: 4.3, Packaging Group: I
    Health Hazard 2 Do not breathe dust. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Density 2.51g/cm3
    EINECS Number 215-142-0
    Molecular Formula Ca3P2
    MDL Number MFCD00015986
    CAS 1305-99-3
    Sensitivity Moisture sensitive
    Health Hazard 1 H260-H300-H400: In contact with water releases flammable gases which may ignite spontaneously. Fatal if swallowed. Very toxic to aquatic life.
    Chemical Name or Material Calcium phosphide
    Grade Technical Grade

    Do not breathe dust. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H260-H300-H400: In contact with water releases flammable gases which may ignite spontaneously. Fatal if swallowed. Very toxic to aquatic life.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit