Alfa Aesar™ Calcium pyruvate, anhydrous, 97%

Manufacturer: Alfa Aesar™ B2520436

Quantity 500g

 View more versions of this product

Catalog No. AAB2520436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52009-14-0
    Molecular Formula C6H6CaO6
    Formula Weight 214.19
    MDL Number MFCD00037199
    Synonym Pyruvic acid calcium salt, anhydrous
    Chemical Name or Material Calcium pyruvate, anhydrous

    Specifications

    Formula Weight 214.19
    Molecular Formula C6H6CaO6
    MDL Number MFCD00037199
    CAS 52009-14-0
    Synonym Pyruvic acid calcium salt, anhydrous
    Quantity 500g
    Chemical Name or Material Calcium pyruvate, anhydrous
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit