Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Calcium pyruvate, anhydrous, 97%

Click to view available options
Quantity:
25g
100g
500g
52009-14-0
C6H6CaO6
214.186
MFCD00037199
UZWMCCLZMHPPKW-UHFFFAOYSA-L
calcium pyruvate,calcium 2-oxopropanoate,unii-9v2cqz19n4,2-oxo-propanoic acid calcium salt,calcium pyruvate, anhydrous,propanoic acid, 2-oxo-, calcium salt,pyruvic acid calcium salt,calcium pyruvic,acmc-20ali9,calcium dipyruvate
9859191
calcium;2-oxopropanoate
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2]
52009-14-0
C6H6CaO6
214.186
MFCD00037199
UZWMCCLZMHPPKW-UHFFFAOYSA-L
calcium pyruvate,calcium 2-oxopropanoate,unii-9v2cqz19n4,2-oxo-propanoic acid calcium salt,calcium pyruvate, anhydrous,propanoic acid, 2-oxo-, calcium salt,pyruvic acid calcium salt,calcium pyruvic,acmc-20ali9,calcium dipyruvate
9859191
calcium;2-oxopropanoate
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2]
Quantity 100g
Formula Weight 214.19
Percent Purity 97%
Chemical Name or Material Calcium pyruvate, anhydrous