Thermo Scientific™ Calcium Reagents
Determine in quantity of calcium in serum or plasma in vitro with Thermo Scientific™ Calcium Reagents.
 View more versions of this product
Catalog No. 1265250
  Check Availability
Add to Cart
    Specifications
    Description Infinity Calcium Arsenazo III
    Methods Compatibility Endpoint
    Storage Requirements 2° to 8°C
    Linearity 6 to 15mg/dL
    Stability Stable Liquid
    State at Room Temperature Liquid
    CE Marker Yes
    Certifications/Compliance CE Marked
    Item Description Infinity Calcium Arsenazo III 2 x 125mL
    Determine in quantity of calcium in serum or plasma in vitro with Thermo Scientific™ Calcium Reagents.
    Calcium plays an essential role in bone mineralization, blood coagulation, muscle contraction and relaxation, nerve signaling and release of certain hormones. Increased levels are associated with hyperparathyroidism, metastatic bone lesions, and Vitamin A or D overdose. Decreased levels are most commonly associated with hypoalbuminemia but can also be observed in cases of hypoparathyroidism, Vitamin D deficiency or resistance, chronic renal failure and calcium losing syndromes. Reagent has up to 18 months of stability from the date of manufacture.
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback