Learn More
Thermo Scientific™ Calcium Reagents
Determine in quantity of calcium in serum or plasma in vitro with Thermo Scientific™ Calcium Reagents.
$217.00
Specifications
Methods Compatibility Endpoint
Stability Stable Liquid
State at Room Temperature Liquid
View More Specs
Products 1
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
1265250
Accessories View Documents
Infinity Calcium Arsenazo III
Pack of 2 for $217.00
Add to cart
Description
Description
Determine in quantity of calcium in serum or plasma in vitro with Thermo Scientific™ Calcium Reagents.
Calcium plays an essential role in bone mineralization, blood coagulation, muscle contraction and relaxation, nerve signaling and release of certain hormones. Increased levels are associated with hyperparathyroidism, metastatic bone lesions, and Vitamin A or D overdose. Decreased levels are most commonly associated with hypoalbuminemia but can also be observed in cases of hypoparathyroidism, Vitamin D deficiency or resistance, chronic renal failure and calcium losing syndromes. Reagent has up to 18 months of stability from the date of manufacture.
Specifications
Specifications
Endpoint
Stable Liquid
Liquid