Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Saccharate, Tetrahydrate, USP, 98.5-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5793-89-5
C6H16CaO12
320.26
ZQWFSIZRQANUDA-FARGFPGJSA-L
calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O
5793-89-5
C6H16CaO12
320.26
ZQWFSIZRQANUDA-FARGFPGJSA-L
calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O

C1830, 5793-89-5, C6H8CaO8•4H2O

Spectrum™ Calcium Saccharate, Tetrahydrate, USP is used as an antacid, as an antidote in carbolic acid poisoning, and as a stabilizer for calcium gluconate solution for parenteral administration. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98.5-102%
Chloride 0.07%
Packaging Amber Glass Bottle
Specific Rotation +18.5° to +22.5°
Sulfate 0.12%
Quantity 100 g