Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Saccharin, FCC, 98-101%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6381-91-5
C14H10CaN2O7S2
422.44
XOCXGVIOTKFUSU-UHFFFAOYSA-L
calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12
6381-91-5
C14H10CaN2O7S2
422.44
XOCXGVIOTKFUSU-UHFFFAOYSA-L
calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12

C1146, 6381-91-5, C14H8CaN2O6S2•31/2H2O

Spectrum™ Calcium Saccharin, FCC is an artificial sweetener that is about 300 times as sweet as table sugar (sucrose). Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98-101%
Lead (Pb) 2 mg/kg
Packaging Poly Bottle
Selenium (Se) 0.003%
Water 15%
Quantity 125 g