Learn More
Calcium Saccharin, FCC, 98-101%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
C1146, 6381-91-5, C14H8CaN2O6S2•31/2H2O
Spectrum™ Calcium Saccharin, FCC is an artificial sweetener that is about 300 times as sweet as table sugar (sucrose). Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Selenium (Se) 3e-005
Water 0.15