Learn More
Calcium-sensing R/CaSR Mouse anti-Human, Alexa Fluor™ 488, Clone: 611825, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9608G100UG
Catalog No. FAB9608G100
Add to cart
This item is not returnable. View return policy
Description
Description
Calcium-sensing R/CaSR Monoclonal antibody specifically detects Calcium-sensing R/CaSR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Calcium-sensing R/CaSR
611825
Flow Cytometry 0.25-1 μg/106 cells
P41180-1
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2A, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Mouse myeloma cell line NS0-derived transfected with recombinant human Calcium-sensing R/CaSR protein, Tyr20-Lys601, Accession # P41180-1
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Calcium-sensing R/CaSR protein in direct ELISAs.
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2a
100 μg
Primary
846
Human