Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium-sensing R/CaSR Mouse anti-Human, Alexa Fluor™ 700, Clone: 611825, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9608N100UG
This item is not returnable.
View return policy
Description
Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human samples. It is validated for Flow Cytometry.Specifications
Calcium-sensing R/CaSR | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2A, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441 | |
Mouse myeloma cell line NS0-derived transfected with recombinant human Calcium-sensing R/CaSR protein, Tyr20-Lys601, Accession # P41180-1 | |
100 μg | |
846 | |
Human | |
Supernatant |
Flow Cytometry | |
611825 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P41180-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Calcium-sensing R/CaSR protein in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title