Alfa Aesar™ Calcium shot, 1cm (0.4 in.) & down, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1012636

Quantity 500g

 View more versions of this product

Catalog No. AA1012636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-70-2
    MDL Number MFCD00085314
    UN Number UN1401
    Chemical Name or Material Calcium shot

    Specifications

    Health Hazard 3 GHS P Statement
    P231+P232-P280-P233-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H261
    In contact with water releases flammable gas.
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00085314
    CAS 7440-70-2
    UN Number UN1401
    Health Hazard 1 Danger
    Physical Form Shot
    Particle Size 1cm (0.4 in.) & down
    Color Metallic
    RTECS Number EV8040000
    Chemical Name or Material Calcium shot
    Assay 99%, (metals basis)

    GHS H Statement
    H261
    In contact with water releases flammable gas.

    GHS P Statement
    P231+P232-P280-P233-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit