Calcium shot, 99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1012795

Quantity 2 x 500g

 View more versions of this product

Catalog No. AA1012795

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-70-2
    Physical Form Shot, Redistilled, 1cm (0.4 in.) &and down
    Assay Percent Range 99.5% (Metals basis)
    MDL Number MFCD00085314
    Chemical Name or Material Calcium

    Specifications

    Physical Form Shot, Redistilled, 1cm (0.4 in.) &and down
    Quantity 2 x 500g
    Color Silver-white
    DOT Information DOT Class: 4.3, Packaging Group: II
    EINECS Number 213-179-5
    Assay Percent Range 99.5% (Metals basis)
    MDL Number MFCD00085314
    CAS 7440-70-2
    Odor Odorless
    Vapor Pressure 0 hPa
    Health Hazard 1 H261: In contact with water releases flammable gas.
    Appearance Silvery-white shot
    Chemical Name or Material Calcium

    H261: In contact with water releases flammable gas.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit