Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium shot, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1012730

Quantity 250g

 View more versions of this product

Catalog No. AA1012730


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-70-2
99.5% (Metals basis)
Calcium
Shot, Redistilled, 1cm (0.4 in.) &and down
MFCD00085314

Specifications

250g
Silvery-white shot
99.5% (Metals basis)
Odorless
Silver-white
7440-70-2
Shot, Redistilled, 1cm (0.4 in.) &and down
MFCD00085314
0 hPa
Calcium

Safety and Handling

SDS & Safety Documents

H261: In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 213-179-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit