Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium silicate, meta, 99% (metals basis), Alfa Aesar

Manufacturer:  Alfa Aesar 3584322

Quantity 100g

 View more versions of this product

Catalog No. AA3584322


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10101-39-0
99%
116.16
Calcium silicate, meta
Powder
CaSiO3
MFCD00015979

Specifications

10101-39-0
99%
116.16
MFCD00015979
Odorless
100g
Powder
CaSiO3
White powder
2.9g/cm3 at 20°C
White
Calcium silicate, meta

Safety and Handling

SDS & Safety Documents

H319-H335 : Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 233-250-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit