Calcium silicate, meta, 99% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 3584314

Quantity 25g

 View more versions of this product

Catalog No. AA3584314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10101-39-0
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula CaSiO3
    Formula Weight 116.16
    MDL Number MFCD00015979
    Chemical Name or Material Calcium silicate, meta

    Specifications

    Formula Weight 116.16
    Quantity 25g
    Color White
    Density 2.9g/cm3 at 20°C
    EINECS Number 233-250-6
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99%
    Appearance White powder
    MDL Number MFCD00015979
    CAS 10101-39-0
    Health Hazard 1 H319-H335 : Causes serious eye irritation. May cause respiratory irritation.
    Molecular Formula CaSiO3
    Chemical Name or Material Calcium silicate, meta

    H319-H335 : Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit