Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Calcium silicide, tech. Ca »30%, may contain up to 5% Fe

Manufacturer:  Alfa Aesar™ 1467609

Quantity 10g

 View more versions of this product

Catalog No. AA1467609


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12013-56-8
MFCD00015977
Calcium silicide
96.25
UN1405
Technical Grade

Specifications

12013-56-8
96.25
UN1405
Ca ≈30%, may contain up to 5% Fe
Moisture Sensitive
≈1000°C
Calcium silicide
Powder
MFCD00015977
Insoluble in cold water, decomposes in hot water
2.5g/mL
Gray
10g
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261
In contact with water releases flammable gas.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM SILICIDE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit